Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-TNFA Rabbit Monoclonal Antibody (Unconjugated) [clone: IPO-38]
Catalog # ROCK110-401-321
Anti-TNFA Rabbit Monoclonal Antibody (Unconjugated) [clone: IPO-38]
Catalog # ROCK110-401-321

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tumor necrosis factor
  • Antigen symbol:
    TNF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    21926
  • Antigen synonyms:
    DIF|Tnfa|Tnfsf1a|TNFalpha|TNF-alpha|TNF-a|TNFSF2
  • Storage temperature:
    Cool
  • Concentration:
    70 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    The whole rabbit serum was prepared by repeated immunizations with recombinant mouse TNFa produced in E.coli.
  • Tested applications:
    Radio Immunoassay
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Primary Rabbit Anti-Mouse TNF alpha Reacts with Mouse

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: IPO-38
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse