Anti-RFX5 Rabbit Monoclonal Antibody (Unconjugated) [clone: BP53-12]
ROCK100-401-194
- Antibody type:Primary
- Antigen name:Regulatory factor X, 5 (influences HLA class II expression)
- Antigen symbol:RFX5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:5993
- Amino acid number:320 to 494
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:85 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:RFX5 peptide corresponding to amino acids 320 to 494 of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
Anti-RFX5 Antibody was also tested in a gel supershift assay and found to be reactive against RFX5 complexes using 0.5 to 1.0 µl per assay. Specific conditions should be optimized by user.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: RFX5
Clonality: Monoclonal
Clone: BP53-12
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human