Anti-c-ErbB2/c-Neu (Ab-5) (TA-1) Mouse Monoclonal Antibody [clone: TA-1]
Catalog # OP39-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- Antigen symbol:C-ERBB2
- Clonality:Monoclonal
- Clone:TA-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-5) (TA-1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human