Anti-5-Methylcytosine (162 33 D3) Mouse Monoclonal Antibody
NA81-50
- Antibody type:Primary
- Antigen name:5-Methylcytosine
- Antigen symbol:5-mC
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Multiple
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: 5-Methylcytosine (162 33 D3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan