Order Entry
Netherlands
ContactUsLinkComponent
Anti-c-Myc tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-c-Myc tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # GENSA00173-40
Supplier:  Genscript
undefined
Anti-c-Myc tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # GENSA00173-40
Supplier:  Genscript

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    EQKLISEEDL
  • Antigen symbol:
    c-Myc tag
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    40 µg
  • Pk:
    40 µG

Specifications

About this item

Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: