Anti-PCK Mouse Monoclonal Antibody [clone: IML-91]
Catalog # BSBTMA1081
Leverancier: Boster Bio
Specificaties
- Antibody type:Primary
- Antigen symbol:PCK
- Clonality:Monoclonal
- Clone:IML-91
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:mixture of several monoclonal cytokeratin clones.
- Size:100 µg
- Pk:0,1 mg
Specificaties
Over dit artikel
Mouse IgG monoclonal antibody for PCK detection. Tested with WB, IHC-P, IHC-F in Human; rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: PCK
Clonality: Monoclonal
Clone: IML-91
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a/ IgG1
Reactivity: Human, Rat