Anti-DACH1 Rabbit Polyclonal Antibody
NOVUNBP2-27192
- Antibody type:Primary
- Antigen name:dachshund family transcription factor 1
- Antigen symbol:DACH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:1602
- Antigen synonyms:DACH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: DACH1 (dachshund family transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: