Anti-VLDLR Mouse Monoclonal Antibody (DyLight® 680) [clone: 6A6]
Catalog # NOVUNBP1-78162FR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Very low density lipoprotein receptor
- Antigen symbol:VLDLR
- Clonality:Monoclonal
- Clone:6A6
- Conjugation:DyLight® 680
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Gene ID:7436
- Antigen synonyms:CAMRQ1|VLDLRCH|CHRMQ1|CARMQ1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: VLDLR (very low density lipoprotein receptor)
Clonality: monoclonal
Clone: 6A6
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat