Anti-RBFOX3/NeuN Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-77686V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:RNA binding protein, fox-1 homolog (C. elegans) 3
- Antigen symbol:RBFOX3
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Gene ID:146713
- Antigen synonyms:FOX-3|FOX3|NEUN|HRNBP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: RBFOX3/NeuN
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat