Anti-Lamin A + C R453W Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 12A-2F5]
NOVUNBP1-77401F
- Antibody type:Primary
- Antigen name:lamin A/C
- Antigen symbol:LMNA
- Clonality:Monoclonal
- Clone:12A-2F5
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:4000
- Antigen synonyms:LMNL1|FPLD|FPL|IDC|PRO1|LMNC|CMT2B1|LGMD1B|LMN1|CDCD1|LFP|FPLD2|LDP1|CDDC|CMD1A|EMD2|HGPS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: Lamin A + C R453W
Clonality: monoclonal
Clone: 12A-2F5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human