Anti-MFN1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 11E9-1H12]
Catalog # NOVUNBP1-71775C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Mitofusin 1
- Antigen symbol:MFN1
- Clonality:Monoclonal
- Clone:11E9-1H12
- Conjugation:DyLight® 650
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:55669
- Antigen synonyms:hfzo1|hfzo2|MFN1|Fzo homolog|Mitofusin-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: MFN1 (mitofusin 1)
Clonality: monoclonal
Clone: 11E9-1H12
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat