Anti-TEK Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNBP1-69753V
- Antibody type:Primary
- Antigen name:TEK tyrosine kinase, endothelial
- Antigen symbol:TEK
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:7010
- Antigen synonyms:TIE-2|VMCM|TIE2|CD202B|VMCM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse