Order Entry
Netherlands
ContactUsLinkComponent
Anti-NOS3 Mouse Monoclonal Antibody [clone: 6H2]
Anti-NOS3 Mouse Monoclonal Antibody [clone: 6H2]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-NOS3 Mouse Monoclonal Antibody [clone: 6H2]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-51582SS
  • NOVUNBP1-51582
  • Antigen symbol:
  • NOS3
  • ENOS
  • Antigen name:
  • Nitric Oxide Synthase 3
  • Endothelial Nitric Oxide Synthase
  • Antigen synonyms:
  • eNOS|ECNOS
  • eNOS|nitric oxide synthase 3 endothelial cell|ECNOS|NOS3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 6H2
  • 6H2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4846
  • 4846
  • Isotype:
  • IgG1
  • IgG1
  • ELISA:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together