Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]

Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]
 
Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]
Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]

 

 

Type: Primary
Antigen: MAP1B (microtubule-associated protein 1B)
Clonality: polyclonal
Clone: SuperBUGS
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:

 

 

  • Catalog No:
  • NOVUNBP1-42827
  • NOVUNBP1-42827SS
  • Antigen symbol:
  • MAP1B
  • MAP1B
  • Antigen name:
  • Microtubule-associated protein 1B
  • Microtubule-associated protein 1B
  • Antigen synonyms:
  • MAP-1B|MAP5|PPP1R102|Microtubule-associated protein 1B|MAP1B|FUTSCH
  • MAP-1B|MAP5|PPP1R102|Microtubule-associated protein 1B|MAP1B|FUTSCH
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • SuperBUGS
  • SuperBUGS
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4131
  • 4131
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Product Family Options

 

  • Size
     
     
  • 100 µL
       
       
    1 * 0,1 mL
    1 * 0,1 mL - 606,00 €
    606,00 €
    1 * 0,1 mL
       606,00 €

     
     
    Specifications
    Cat. No.
    NOVUNBP1-42827
    Antigen symbol
    MAP1B
    Antigen name
    Microtubule-associated protein 1B
    Antigen synonyms
    MAP-1B|MAP5|PPP1R102|Microtubule-associated protein 1B|MAP1B|FUTSCH
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Clone
    SuperBUGS
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    4131
    Western blot
    Yes
    ImmunoFluorescence
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • 25 µL
       
       
    1 * 25 µl
    1 * 25 µl - 255,00 €
    255,00 €
    1 * 25 µl
       255,00 €

     
     
    Specifications
    Cat. No.
    NOVUNBP1-42827SS
    Antigen symbol
    MAP1B
    Antigen name
    Microtubule-associated protein 1B
    Antigen synonyms
    MAP-1B|MAP5|PPP1R102|Microtubule-associated protein 1B|MAP1B|FUTSCH
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Clone
    SuperBUGS
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    4131
    Western blot
    Yes
    ImmunoFluorescence
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice