Order Entry
Netherlands
ContactUsLinkComponent
Anti-HIST4H4 Mouse Monoclonal Antibody (Biotin) [clone: 5E10-D8]
Anti-HIST4H4 Mouse Monoclonal Antibody (Biotin) [clone: 5E10-D8]
Catalog # NOVUNBP1-30091B
Supplier:  Novus Biologicals
Anti-HIST4H4 Mouse Monoclonal Antibody (Biotin) [clone: 5E10-D8]
Catalog # NOVUNBP1-30091B
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Histone cluster 4 H4
  • Antigen symbol:
    HIST4H4
  • Clonality:
    Monoclonal
  • Clone:
    5E10-D8
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    121504
  • Antigen synonyms:
    H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|H4/G|HIST2H4B|H4F2|H4/O|HIST1H4L|H4/D|H4FC|H4/E|H4/B|H4FE|HIST1H4J|H4FH|H4FM|H4FO|H4FB|HIST1H4K|H4/N|H4/p|H4/J|HIST2H4|H4/C|HIST1H4F|H4FG|HIST1H4I|H4/A|H4FK|H4/H|H4FN|H4FD|HIST2H4A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: monoclonal
Clone: 5E10-D8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: