Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 405) [clone: OC2-3C7]
NOVUNBP1-18970V
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:OC2-3C7
- Conjugation:DyLight® 405
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-3C7
Conjugation: DyLight® 405
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse