Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 680) [clone: OC2-3C7]
Catalog # NOVUNBP1-18970FR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:OC2-3C7
- Conjugation:DyLight® 680
- Flow cytometry:Yes
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-3C7
Conjugation: DyLight® 680
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse