Order Entry
Netherlands
ContactUsLinkComponent
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 650) [clone: OC2-3C7]
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 650) [clone: OC2-3C7]
Catalog # NOVUNBP1-18970C
Supplier:  Novus Biologicals
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 650) [clone: OC2-3C7]
Catalog # NOVUNBP1-18970C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Oval Cell Marker
  • Clonality:
    Monoclonal
  • Clone:
    OC2-3C7
  • Conjugation:
    DyLight® 650
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-3C7
Conjugation: DyLight® 650
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse