Anti-HPd2 Mouse Monoclonal Antibody [clone: DHIC2-5H10]
Type: Primary
Antigen: HPd2
Clonality: monoclonal
Clone: DHIC2-5H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
- Catalog No:
- NOVUNBP1-18955
- NOVUNBP1-18955SS
- Antigen symbol:
- HPD2
- HPD2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DHIC2-5H10
- DHIC2-5H10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgM
- IgM
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together