Order Entry
Netherlands
ContactUsLinkComponent
Anti-HPx2 Mouse Monoclonal Antibody [clone: HIC0-1C10]
Anti-HPx2 Mouse Monoclonal Antibody [clone: HIC0-1C10]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-HPx2 Mouse Monoclonal Antibody [clone: HIC0-1C10]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-18952SS
  • NOVUNBP1-18952
  • Antigen symbol:
  • HPx2
  • HPx2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • HIC0-1C10
  • HIC0-1C10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgM
  • IgM
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together