Anti-HPx1 Mouse Monoclonal Antibody (DyLight® 680) [clone: HIC0-3B3]
Catalog # NOVUNBP1-18951FR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:HPX1
- Clonality:Monoclonal
- Clone:HIC0-3B3
- Conjugation:DyLight® 680
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: HPx1
Clonality: monoclonal
Clone: HIC0-3B3
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human