Anti-HPSE2 Mouse Monoclonal Antibody (DyLight® 680) [clone: DHIC2-2B4]
Catalog # NOVUNBP1-18950FR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:heparanase 2
- Antigen symbol:HPSE2
- Clonality:Monoclonal
- Clone:DHIC2-2B4
- Conjugation:DyLight® 680
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: HPSE2 (heparanase 2)
Clonality: monoclonal
Clone: DHIC2-2B4
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human