Anti-CCL2 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Specifications
- Catalog No:
- NOVUNBP1-07034
- NOVUNBP1-07034SS
- Antigen symbol:
- MCP1
- MCP1
- Antigen name:
- Monocyte Chemoattractant Protein 1
- Monocyte Chemoattractant Protein 1
- Antigen synonyms:
- monocyte chemoattractant protein-1|small inducible cytokine A2 (monocyte chemotactic protein 1|small inducible cytokine subfamily A (Cys-Cys)|monocyte secretory protein JE|monocyte chemotactic and activating factor|member 2|monocyte chemotactic protein 1|homologous to mouse Sig-je)|small-inducible cytokine A2|monocyte chemoattractant protein 1|C-C motif chemokine 2
- monocyte chemoattractant protein-1|small inducible cytokine A2 (monocyte chemotactic protein 1|small inducible cytokine subfamily A (Cys-Cys)|monocyte secretory protein JE|monocyte chemotactic and activating factor|member 2|monocyte chemotactic protein 1|homologous to mouse Sig-je)|small-inducible cytokine A2|monocyte chemoattractant protein 1|C-C motif chemokine 2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6347
- 6347
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together