Anti-NBS1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-06609G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Nibrin
- Antigen symbol:NBN
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:4683
- Antigen synonyms:P95|AT-V1|NBS|AT-V2|FLJ10155|MGC87362|NBS1|ATV
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: NBS1
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse