Order Entry
Netherlands
ContactUsLinkComponent
Anti-NBS1 Rabbit Polyclonal Antibody
Anti-NBS1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-NBS1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-06609SS
  • NOVUNBP1-06609
  • Antigen symbol:
  • NBN
  • NBN
  • Antigen name:
  • Nibrin
  • Nibrin
  • Antigen synonyms:
  • P95|AT-V1|NBS|AT-V2|FLJ10155|MGC87362|NBS1|ATV
  • P95|AT-V1|NBS|AT-V2|FLJ10155|MGC87362|NBS1|ATV
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4683
  • 4683
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together