Anti-HSPA5 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Specifications
- Catalog No:
- NOVUNBP1-06277SS
- NOVUNBP1-06277
- Antigen symbol:
- GRP78
- GRP78
- Antigen name:
- Glucose-regulated protein 78kDa
- Glucose-regulated protein 78kDa
- Antigen synonyms:
- GRP78|BIP|MIF2|HEL-S-89n
- GRP78|BIP|MIF2|HEL-S-89n
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 3309
- 3309
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together