Order Entry
Netherlands
ContactUsLinkComponent
Anti-Aflatoxin B1 Mouse Monoclonal Antibody [clone: 6A10]
Anti-Aflatoxin B1 Mouse Monoclonal Antibody [clone: 6A10]
Supplier:  Novus Biologicals
Certificates
Anti-Aflatoxin B1 Mouse Monoclonal Antibody [clone: 6A10]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB600-443
  • NOVUNB600-443SS
  • Antigen symbol:
  • AFB1
  • AFB1
  • Antigen name:
  • Aflatoxin B1
  • Aflatoxin B1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 6A10
  • 6A10
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together