Order Entry
Netherlands
ContactUsLinkComponent
Anti-BECN1 Rabbit Polyclonal Antibody
Anti-BECN1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-BECN1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB500-249SS
  • NOVUNB500-249
  • Antigen symbol:
  • BECN1
  • BECN1
  • Antigen name:
  • Beclin 1
  • Beclin 1
  • Antigen synonyms:
  • Coiled-coil myosin-like BCL2-interacting protein|APG 6|VPS30|Beclin1 autophagy|ATG6|CTIP-1 COUP-TF interacting protein 1|Protein GT197
  • Coiled-coil myosin-like BCL2-interacting protein|APG 6|VPS30|Beclin1 autophagy|ATG6|CTIP-1 COUP-TF interacting protein 1|Protein GT197
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 8678
  • 8678
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together