Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CASP9 Mouse monoclonal antibody unconjugated [clone: LAP6 96-2-22]
 
undefined
Anti-CASP9 Mouse monoclonal antibody unconjugated [clone: LAP6 96-2-22]

 

  • Catalog No:
  • NOVUNB500-209SS
  • NOVUNB500-209
  • Antigen symbol:
  • CASP9
  • CASP9
  • Antigen name:
  • Caspase 9
  • Caspase 9
  • Antigen synonyms:
  • EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
  • EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • LAP6 96-2-22
  • LAP6 96-2-22
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 842
  • 842
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together