Anti-SSTR1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: SSTR1 (somatostatin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNB300-120
- NOVUNB300-120SS
- Antigen symbol:
- SSTR1
- SSTR1
- Antigen name:
- Somatostatin Receptor 1
- Somatostatin Receptor 1
- Antigen synonyms:
- Somatostatin receptor 1|SS1-R|SRIF-2|SS-1-R|SS1R
- Somatostatin receptor 1|SS1-R|SRIF-2|SS-1-R|SS1R
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6751
- 6751
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together