Order Entry
Netherlands
ContactUsLinkComponent
Anti-MMP2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 8B4]
Anti-MMP2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 8B4]
Catalog # NOVUNB200-114F
Supplier:  Novus Biologicals
Anti-MMP2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 8B4]
Catalog # NOVUNB200-114F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
  • Antigen symbol:
    MMP2
  • Clonality:
    Monoclonal
  • Clone:
    8B4
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    4313
  • Antigen synonyms:
    MMP-2|MMP-II|MONA|CLG4A|CLG4|TBE-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: monoclonal
Clone: 8B4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat