Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB110-93479R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase E1-alpha Subunit
- Antigen symbol:PDC E1A
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:5160
- Antigen synonyms:PDHCE1A|PDHAD|PHE1A|PDHA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha subunit [p Ser293]
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat