Order Entry
Netherlands
ContactUsLinkComponent
Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB110-93479H
Supplier:  Novus Biologicals
Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB110-93479H
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Pyruvate Dehydrogenase E1-alpha Subunit
  • Antigen symbol:
    PDC E1A
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    5160
  • Antigen synonyms:
    PDHCE1A|PDHAD|PHE1A|PDHA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µL
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha subunit [p Ser293]
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat