Anti-Capicua Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB110-59905G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Capicua
- Antigen symbol:CIC
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Gene ID:23152
- Antigen synonyms:KIAA0306|Protein capicua homolog
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Capicua
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: