Anti-NOX4 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: NOX4 (NADPH oxidase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNB110-58849SS
- NOVUNB110-58849
- Antigen symbol:
- NOX4
- NOX4
- Antigen name:
- NAPDH oxidase 4
- NAPDH oxidase 4
- Antigen synonyms:
- NAPDH oxidase 4|KOX-1|KOX|NOX-4|RENOX
- NAPDH oxidase 4|KOX-1|KOX|NOX-4|RENOX
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 50507
- 50507
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together