Order Entry
Netherlands
ContactUsLinkComponent
Anti-CITED4 Mouse Monoclonal Antibody (DyLight® 550) [clone: HT13-2D6.3]
Anti-CITED4 Mouse Monoclonal Antibody (DyLight® 550) [clone: HT13-2D6.3]
Catalog # NOVUNB110-41572R
Supplier:  Novus Biologicals
Anti-CITED4 Mouse Monoclonal Antibody (DyLight® 550) [clone: HT13-2D6.3]
Catalog # NOVUNB110-41572R
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal Domain, 4
  • Antigen symbol:
    CITED4
  • Clonality:
    Monoclonal
  • Clone:
    HT13-2D6.3
  • Conjugation:
    DyLight® 550
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    163732
  • Antigen synonyms:
    cbp/p300-interacting transactivator 4|MRG-2|transcriptional co-activator 4|MSG1-related protein 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: CITED4 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4)
Clonality: monoclonal
Clone: HT13-2D6.3
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human