Anti-LIM1 Rabbit Polyclonal Antibody
NOVUNB100-56626
- Antibody type:Primary
- Antigen name:LIM homeobox 1
- Antigen symbol:LHX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:3975
- Antigen synonyms:LIM-1|LIM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 μL
- Pk:0,2 mL
Type: Primary
Antigen: LIM1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human