Anti-CCR3 Rabbit Polyclonal Antibody
NOVUNB100-56335
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) receptor 3
- Antigen symbol:CCR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:1232
- Antigen synonyms:CD193|CC-CKR-3|CKR3|CMKBR3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: CCR3 (chemokine (C-C motif) receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat