Anti-ZDHHC23 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56149
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, DHHC-type containing 23
- Antigen symbol:ZDHHC23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:254887
- Antigen synonyms:NIDD
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ZDHHC23 (zinc finger, DHHC-type containing 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human