Anti-CFLAR Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56141
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:CASP8 and FADD-like apoptosis regulator
- Antigen symbol:CFLAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:8837
- Antigen synonyms:c-FLIP|c-FLIPS|CLARP|FLAME-1|I-FLICE|MRIT|CASP8AP1|FLAME|c-FLIPL|c-FLIPR|FLIP|FLAME1|CASH|Casper
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CFLAR (CASP8 and FADD-like apoptosis regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human