Anti-FANCD2 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNB100-182
- NOVUNB100-182SS
- Antigen symbol:
- FANCD2
- FANCD2
- Antigen name:
- Fanconi Anemia Complementation Group D2
- Fanconi Anemia Complementation Group D2
- Antigen synonyms:
- FAD|FA4|FANCD|FACD|FA-D2|FAD2
- FAD|FA4|FANCD|FACD|FA-D2|FAD2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2177
- 2177
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together