Order Entry
Netherlands
ContactUsLinkComponent
Anti-RAD51C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H11/6]
Anti-RAD51C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H11/6]
Catalog # NOVUNB100-177PE
Supplier:  Novus Biologicals
Anti-RAD51C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H11/6]
Catalog # NOVUNB100-177PE
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RAD51 paralog C
  • Antigen symbol:
    RAD51C
  • Clonality:
    Monoclonal
  • Clone:
    2H11/6
  • Conjugation:
    PE (Phycoerythrin)
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Gene ID:
    5889
  • Antigen synonyms:
    BROVCA3|RAD51L2|FANCO|R51H3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: RAD51C (RAD51 paralog C)
Clonality: monoclonal
Clone: 2H11/6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: