Anti-XRCC3 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB100-165V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:X-ray repair complementing defective repair in Chinese hamster cells 3
- Antigen symbol:XRCC3
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:7517
- Antigen synonyms:CMM6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human