Anti-Parathyroid Hormone Receptor 1 Rabbit Polyclonal Antibody
Catalog # BSBTPA2132
Leverancier: Boster Bio
Specificaties
- Antibody type:Primary
- Antigen name:Parathyroid Hormone Receptor 1
- Antigen symbol:PTH1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:388 - 406
- Sequence:KLRETNAGRCDTRQQYRKL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Parathyroid Hormone Receptor 1 (388-406aa KLRETNAGRCDTRQQYRKL), identical to the related mouse and rat sequences
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specificaties
Over dit artikel
Rabbit IgG polyclonal antibody for Parathyroid hormone/parathyroid hormone-related peptide receptor(PTH1R) detection. Tested with WB, IHC-P, ICC in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Parathyroid Hormone Receptor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat