Order Entry
Netherlands
ContactUsLinkComponent
Anti-BMPR1B Rabbit Polyclonal Antibody
Anti-BMPR1B Rabbit Polyclonal Antibody
Catalog # BSBTPA2017
Supplier:  Boster Bio
undefined
Anti-BMPR1B Rabbit Polyclonal Antibody
Catalog # BSBTPA2017
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bone morphogenetic Protein Receptor, type IB
  • Antigen symbol:
    BMPR1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    Acvrlk6|BMPR-1B|ALK-6|AI385617|SKR6|AV355320|CFK-43a|CD293|CDw293|Alk6|BMPR-IB
  • Amino acid number:
    145 - 160
  • Sequence:
    FCYFRYKRQETRPRYS
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human BMPR1B (145-160aa FCYFRYKRQETRPRYS), different from the related rat and mouse sequences by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Bone morphogenetic protein receptor type-1B (BMPR1B) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: BMPR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat