Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CaMKII alpha (phospho T286) Rabbit Monoclonal Antibody [clone: EPR1830]
 :  Abcam
New Product
 
undefined
Anti-CaMKII alpha (phospho T286) Rabbit Monoclonal Antibody [clone: EPR1830]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB124880-100
  • ABCAAB124880-1ML
  • Antigen symbol:
  • CAMK2A
  • Antigen name:
  • calcium/calmodulin-dependent Protein kinase II alpha
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR1830
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

Frequently Bought Together