Order Entry
Netherlands
ContactUsLinkComponent
Anti-DPP4 Mouse Monoclonal Antibody [clone: TP1/19] (CF® 405M)
Anti-DPP4 Mouse Monoclonal Antibody [clone: TP1/19] (CF® 405M)
Catalog # ABCAAB119480-50
Supplier:  Abcam
undefined
Anti-DPP4 Mouse Monoclonal Antibody [clone: TP1/19] (CF® 405M)
Catalog # ABCAAB119480-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dipeptidylpeptidase 4
  • Antigen symbol:
    DPP4
  • Clonality:
    Monoclonal
  • Clone:
    TP1/19
  • Conjugation:
    CF® 405M
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Seq:
    0780
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 Tests
  • Pack type:
    Vial
  • Pk:
    50 Tests

Specifications

About this item

CF405M Mouse monoclonal [TP1/19] to DPP4.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: TP1/19
Conjugation: CF® 405M
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human