Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-MyD88 Mouse Monoclonal Antibody [clone: OTI1B4]
  ABCAAB119048-100
 :  Abcam
undefined
Anti-MyD88 Mouse Monoclonal Antibody [clone: OTI1B4]
  ABCAAB119048-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Myeloid differentiation primary response 88
  • Antigen symbol:
    MYD88
  • Clonality:
    Monoclonal
  • Clone:
    1B4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1286
  • Form:
    Liquid
  • Antigen synonyms:
    MYD88|Myeloid differentiation primary response 88|Myeloid differentiation primary response protein MyD88
  • Storage buffer:
    pH: 7,3
    Preservative: 0,02% Sodium azide
    Constituents: PBS, 50% Glycerol (glycerin, glycerine), 1% BSA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    WB
    ImmunoChemistry
    Flow cytometry
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Mouse monoclonal [OTI1B4] to MyD88.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MYD88 (myeloid differentiation primary response 88)
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human