Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-Pyruvate Dehydrogenase E2 Mouse Monoclonal Antibody [clone: 15D3G9C11]
Catalog # ABCAAB110332-100
Supplier:  Abcam
undefined
Anti-Pyruvate Dehydrogenase E2 Mouse Monoclonal Antibody [clone: 15D3G9C11]
Catalog # ABCAAB110332-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Pyruvate Dehydrogenase E2
  • Clonality:
    Monoclonal
  • Clone:
    15D3G9C11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [15D3G9C11] to Pyruvate Dehydrogenase E2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Pyruvate Dehydrogenase E2
Clonality: Monoclonal
Clone: 15D3G9C11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human