Order Entry
Netherlands
ContactUsLinkComponent
Anti-PI3 Kinase p110 beta Rabbit Polyclonal Antibody
Anti-PI3 Kinase p110 beta Rabbit Polyclonal Antibody
  ABCAAB97322-50
 :  Abcam
Anti-PI3 Kinase p110 beta Rabbit Polyclonal Antibody
  ABCAAB97322-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    PI3 kinase p110 beta
  • Antigen symbol:
    P110B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    50 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PI3 Kinase p110 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human